SY3H3D67FE43 Synthonix Corporation

Methyl 4H-thieno[3,2-b]pyrrole-5-carboxylate

  • Cas Number : 82782-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/48493.html

Safety Information

MSDS https://synthonix.com/48493.html
Availability details for
Quantity
  • Availability
  • Add to Cart