SY3H3D67FE46 Synthonix Corporation

2-Methyl-5-nitrobenzaldehyde

  • MDL number MFCD03840506
  • Cas Number : 16634-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: Very pale yellow - Reddish yellow or Gray
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48498.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48498.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart