SY3H3D67FE4A Synthonix Corporation

5-Methyl-2-nitrobenzaldehyde

  • MDL number No MDL found
  • Cas Number : 5858-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/48502.html

Safety Information

MSDS https://synthonix.com/48502.html
Availability details for
Quantity
  • Availability
  • Add to Cart