SY3H3D67FE4B Synthonix Corporation

Methyl 2-formylisonicotinate

  • MDL number No MDL found
  • Cas Number : 125104-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/48503.html

Safety Information

MSDS https://synthonix.com/48503.html
Availability details for
Quantity
  • Availability
  • Add to Cart