SY3H3D67FE4D Synthonix Corporation

Methyl 5-bromo-2-chloro-1-methyl-6-oxo-1,6-dihydropyridine-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 869357-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48508.html

Safety Information

MSDS https://synthonix.com/48508.html
Availability details for
Quantity
  • Availability
  • Add to Cart