SY3H3D67FE55 Synthonix Corporation

4-Methoxy-2-nitrobenzaldehyde

  • MDL number MFCD00024254
  • Cas Number : 22996-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Off white to Pale-yellow to Yellow-brown or Orange Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48517.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48517.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart