SY3H3D67FE57 Synthonix Corporation

4-(Methoxycarbonyl)picolinic acid

  • MDL number No MDL found
  • Cas Number : 24195-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48519.html

Safety Information

MSDS https://synthonix.com/48519.html
Availability details for
Quantity
  • Availability
  • Add to Cart