SY3H3D67FE58 Synthonix Corporation

5-Methyl-6-nitrobenzo[d][1,3]dioxole

  • Cas Number : 32996-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48524.html

Safety Information

MSDS https://synthonix.com/48524.html
Availability details for
Quantity
  • Availability
  • Add to Cart