SY3H3D67FE59 Synthonix Corporation

5-Methylpyridine-2,3-dicarboxylic acid

  • MDL number No MDL found
  • Cas Number : 53636-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48525.html

Safety Information

MSDS https://synthonix.com/48525.html
Availability details for
Quantity
  • Availability
  • Add to Cart