SY3H3D67FE64 Synthonix Corporation

Methyl 3-hydroxy-4-nitrobenzoate

  • MDL number MFCD00016997
  • Cas Number : 713-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: Pale yellow - Deep yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48537.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48537.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart