SY3H3D67FE67 Synthonix Corporation

5-(Methylsulfonyl)-2-nitrobenzoic acid

  • Cas Number : 898547-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48545.html

Safety Information

MSDS https://synthonix.com/48545.html
Availability details for
Quantity
  • Availability
  • Add to Cart