SY3H3D67FE68 Synthonix Corporation

6-Methoxybenzo[d]thiazole

  • MDL number No MDL found
  • Cas Number : 2942-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48553.html

Safety Information

MSDS https://synthonix.com/48553.html
Availability details for
Quantity
  • Availability
  • Add to Cart