SY3H3D67FE6A Synthonix Corporation

2-(Methylthio)benzo[d]oxazole

  • MDL number No MDL found
  • Cas Number : 13673-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/48556.html

Safety Information

MSDS https://synthonix.com/48556.html
Availability details for
Quantity
  • Availability
  • Add to Cart