SY3H3D67FE6C Synthonix Corporation

5-Methoxybenzo[d]thiazole-2-thiol

  • MDL number No MDL found
  • Cas Number : 55690-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/48558.html

Safety Information

MSDS https://synthonix.com/48558.html
Availability details for
Quantity
  • Availability
  • Add to Cart