SY3H3D67FE72 Synthonix Corporation

3-Methoxy-5-(trifluoromethyl)phenol

  • MDL number No MDL found
  • Cas Number : 349-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48626.html

Safety Information

MSDS https://synthonix.com/48626.html
Availability details for
Quantity
  • Availability
  • Add to Cart