SY3H3D67FE7C Synthonix Corporation

Methyl 4-chloro-3-hydroxybenzoate

  • Cas Number : 166272-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/48664.html

Safety Information

MSDS https://synthonix.com/48664.html
Availability details for
Quantity
  • Availability
  • Add to Cart