SY3H3D67FE7D Synthonix Corporation

Methyl 2-fluoro-3-hydroxybenzoate

  • MDL number No MDL found
  • Cas Number : 1214324-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48677.html

Safety Information

MSDS https://synthonix.com/48677.html
Availability details for
Quantity
  • Availability
  • Add to Cart