SY3H3D67FE84 Synthonix Corporation

Methyl 5-fluoro-2-hydroxybenzoate

  • Cas Number : 391-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/48684.html

Safety Information

MSDS https://synthonix.com/48684.html
Availability details for
Quantity
  • Availability
  • Add to Cart