SY3H3D67FE92 Synthonix Corporation

8-Methylimidazo[1,2-a]pyridine

  • Cas Number : 874-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48790.html

Safety Information

MSDS https://synthonix.com/48790.html
Availability details for
Quantity
  • Availability
  • Add to Cart