SY3H3D68027D Synthonix Corporation

2-Methyloctahydro-1H-pyrazino[1,2-a]pyrazine trihydrochloride

  • MDL number No MDL found
  • Cas Number : 1883347-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49690.html

Safety Information

MSDS https://synthonix.com/49690.html
Availability details for
Quantity
  • Availability
  • Add to Cart