SY3H3D68027F Synthonix Corporation

5-Methyl-2-aminobenzoxazol

  • MDL number No MDL found
  • Cas Number : 64037-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48797.html

Safety Information

MSDS https://synthonix.com/48797.html
Availability details for
Quantity
  • Availability
  • Add to Cart