SY3H3D680280 Synthonix Corporation

1-Methyl-1H-indazol-3(2H)-one

  • Cas Number : 1006-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48798.html

Safety Information

MSDS https://synthonix.com/48798.html
Availability details for
Quantity
  • Availability
  • Add to Cart