SY3H3D680287 Synthonix Corporation

2-(5-Methoxypyridin-2-yl)acetonitrile

  • Cas Number : 204067-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48814.html

Safety Information

MSDS https://synthonix.com/48814.html
Availability details for
Quantity
  • Availability
  • Add to Cart