SY3H3D680288 Synthonix Corporation

1-Methyl-1H-benzo[d]imidazol-2(3H)-one

  • MDL number MFCD00142654
  • Cas Number : 1849-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48815.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48815.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart