SY3H3D680289 Synthonix Corporation

5-Methyl-1H-benzo[d]imidazol-2(3H)-one

  • MDL number No MDL found
  • Cas Number : 5400-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/48816.html

Safety Information

MSDS https://synthonix.com/48816.html
Availability details for
Quantity
  • Availability
  • Add to Cart