SY3H3D68028A Synthonix Corporation

2-Methyl-1H-pyrrolo[2,3-c]pyridin-7(6H)-one

  • MDL number No MDL found
  • Cas Number : 1427503-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48819.html

Safety Information

MSDS https://synthonix.com/48819.html
Availability details for
Quantity
  • Availability
  • Add to Cart