SY3H3D68028D Synthonix Corporation

5-Methoxy-1H-pyrrolo[3,2-b]pyridin-2(3H)-one

  • Cas Number : 178393-14-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48829.html

Safety Information

MSDS https://synthonix.com/48829.html
Availability details for
Quantity
  • Availability
  • Add to Cart