SY3H3D68028F Synthonix Corporation

Methyl 2-amino-5-nitrobenzoate

  • MDL number No MDL found
  • Cas Number : 3816-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/48839.html

Safety Information

MSDS https://synthonix.com/48839.html
Availability details for
Quantity
  • Availability
  • Add to Cart