SY3H3D680295 Synthonix Corporation

Methyl 5-methoxy-6-nitropicolinate

  • MDL number No MDL found
  • Cas Number : 390816-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48845.html

Safety Information

MSDS https://synthonix.com/48845.html
Availability details for
Quantity
  • Availability
  • Add to Cart