SY3H3D680297 Synthonix Corporation

7-Methylbenzo[d]thiazol-2-amine

  • MDL number No MDL found
  • Cas Number : 14779-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48850.html

Safety Information

MSDS https://synthonix.com/48850.html
Availability details for
Quantity
  • Availability
  • Add to Cart