SY3H3D680299 Synthonix Corporation

Methyl 2,4-diamino-3-fluoro-5-nitrobenzoate

  • MDL number No MDL found
  • Cas Number : 918321-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48858.html

Safety Information

MSDS https://synthonix.com/48858.html
Availability details for
Quantity
  • Availability
  • Add to Cart