SY3H3D68029A Synthonix Corporation

2-(3-Methyl-1H-1,2,4-triazol-5-yl)pyridine

  • MDL number MFCD11106659
  • Cas Number : 25433-36-7
  • Molecular Weight : 160.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/48866.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/48866.html
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart