SY3H3D68029C Synthonix Corporation

3-methyl-5-(trifluoromethyl)aniline

  • MDL number No MDL found
  • Cas Number : 96100-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/48891.html

Safety Information

MSDS https://synthonix.com/48891.html
Availability details for
Quantity
  • Availability
  • Add to Cart