SY3H3D6802A3 Synthonix Corporation

Methyl 4-amino-2-bromobenzoate

  • MDL number No MDL found
  • Cas Number : 98545-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/48911.html

Safety Information

MSDS https://synthonix.com/48911.html
Availability details for
Quantity
  • Availability
  • Add to Cart