SY3H3D6802A9 Synthonix Corporation

Methyl 2-bromo-6-methylnicotinate

  • MDL number No MDL found
  • Cas Number : 885277-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48919.html

Safety Information

MSDS https://synthonix.com/48919.html
Availability details for
Quantity
  • Availability
  • Add to Cart