SY3H3D6802AF Synthonix Corporation

Methyl 4-amino-2-chlorobenzoate

  • Cas Number : 46004-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48941.html

Safety Information

MSDS https://synthonix.com/48941.html
Availability details for
Quantity
  • Availability
  • Add to Cart