SY3H3D6802B6 Synthonix Corporation

Methyl 6-chloro-4-methylpicolinate

  • MDL number No MDL found
  • Cas Number : 1186605-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/48948.html

Safety Information

MSDS https://synthonix.com/48948.html
Availability details for
Quantity
  • Availability
  • Add to Cart