SY3H3D6802BA Synthonix Corporation

Methyl 2-(6-chloropyridin-2-yl)acetate

  • MDL number No MDL found
  • Cas Number : 161807-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48959.html

Safety Information

MSDS https://synthonix.com/48959.html
Availability details for
Quantity
  • Availability
  • Add to Cart