SY3H3D6802C0 Synthonix Corporation

Methyl 3-amino-5-fluorobenzoate

  • MDL number No MDL found
  • Cas Number : 884497-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/48967.html

Safety Information

MSDS https://synthonix.com/48967.html
Availability details for
Quantity
  • Availability
  • Add to Cart