SY3H3D6802C5 Synthonix Corporation

Methyl 3-amino-4-iodobenzoate

  • MDL number MFCD08064225
  • Cas Number : 412947-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow to Brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48978.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48978.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart