SY3H3D6802C7 Synthonix Corporation

Methyl 4-bromo-6-methoxypicolinate

  • Cas Number : 1256789-39-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48986.html

Safety Information

MSDS https://synthonix.com/48986.html
Availability details for
Quantity
  • Availability
  • Add to Cart