SY3H3D6802CA Synthonix Corporation

Methyl 6-bromo-2-methoxynicotinate

  • Cas Number : 1009735-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48989.html

Safety Information

MSDS https://synthonix.com/48989.html
Availability details for
Quantity
  • Availability
  • Add to Cart