SY3H3D6802CF Synthonix Corporation

Methyl 4-chloro-2-methoxynicotinate

  • Cas Number : 1256826-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/49001.html

Safety Information

MSDS https://synthonix.com/49001.html
Availability details for
Quantity
  • Availability
  • Add to Cart