SY3H3D6802DA Synthonix Corporation

N-Methyl-2-(trifluoromethoxy)aniline

  • MDL number No MDL found
  • Cas Number : 175278-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/49060.html

Safety Information

MSDS https://synthonix.com/49060.html
Availability details for
Quantity
  • Availability
  • Add to Cart