SY3H3D6802DE Synthonix Corporation

(3-Methyl-4-(trifluoromethyl)phenyl)boronic acid

  • MDL number No MDL found
  • Cas Number : 864759-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49078.html

Safety Information

MSDS https://synthonix.com/49078.html
Availability details for
Quantity
  • Availability
  • Add to Cart