SY3H3D6802E2 Synthonix Corporation

Methyl 4,6-dihydrothieno[3,4-b]thiophene-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 7767-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49091.html

Safety Information

MSDS https://synthonix.com/49091.html
Availability details for
Quantity
  • Availability
  • Add to Cart