SY3H3D6802E9 Synthonix Corporation

Methyl 2-Furoylacetate

  • Cas Number : 615-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49114.html

Safety Information

MSDS https://synthonix.com/49114.html
Availability details for
Quantity
  • Availability
  • Add to Cart