SY3H3D6802ED Synthonix Corporation

Methyl 2,3-diamino-5-bromobenzoate

  • MDL number No MDL found
  • Cas Number : 1248541-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49177.html

Safety Information

MSDS https://synthonix.com/49177.html
Availability details for
Quantity
  • Availability
  • Add to Cart