SY3H3D6802EF Synthonix Corporation

4-Methyl-1H-indazol-7-amine

  • MDL number No MDL found
  • Cas Number : 952183-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49199.html

Safety Information

MSDS https://synthonix.com/49199.html
Availability details for
Quantity
  • Availability
  • Add to Cart