SY3H3D6802F5 Synthonix Corporation

2-Methyl-2-(pyrimidin-2-yl)propanenitrile

  • MDL number No MDL found
  • Cas Number : 1849313-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49211.html

Safety Information

MSDS https://synthonix.com/49211.html
Availability details for
Quantity
  • Availability
  • Add to Cart